Please login to your online account to display your discounted pricing

(S)-N-FMOC-4-Chlorophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-FMOC-4-Chlorophenylalanine
Name Note 95%, (98% e.e.)
CAS 175453-08-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C24H20ClNO4
Formula Weight 421.87
MDL Number MFCD00080281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC361630010

  SDS
thermo fisher scientific
361630010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 421.87
Physical Form Powder
Quantity 1g
Color Off-white
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 95%
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-chlorophenyl-L-alanine
Name Note 95%, (98% e.e.)
Molecular Formula C24H20ClNO4
CAS 175453-08-4
Chemical Name or Material (S)-N-FMOC-4-Chlorophenylalanine
MDL Number MFCD00080281