Please login to your online account to display your discounted pricing

(S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361560010

 View more versions of this product

Catalog No. AC361560010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystals
Formula Weight 466.33
Quantity 1g
Color White to off-white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
MDL Number MFCD01311760
CAS 220497-68-7
Enantiomeric Excess 97.5% min. (HPLC)
Synonym (S)-N-(9-Fluorenylmethoxycarbonyl)-3-amino-3-(4-bromophenyl)propanoic acid
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material (S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid
Name Note 95%, (98% e.e.)
CAS 220497-68-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
Formula Weight 466.33
MDL Number MFCD01311760
Synonym (S)-N-(9-Fluorenylmethoxycarbonyl)-3-amino-3-(4-bromophenyl)propanoic acid