Please login to your online account to display your discounted pricing

(S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid
Name Note 95%, (98% e.e.)
CAS 220497-68-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
Formula Weight 466.33
MDL Number MFCD01311760
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361560010

  SDS
thermo fisher scientific
361560010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC361565000

  SDS
thermo fisher scientific
361565000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 466.33
Physical Form Crystals
Quantity 1g
Color White to off-white
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Synonym (S)-N-(9-Fluorenylmethoxycarbonyl)-3-amino-3-(4-bromophenyl)propanoic acid
Enantiomeric Excess 97.5% min. (HPLC)
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
Chemical Name or Material (S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid
MDL Number MFCD01311760
CAS 220497-68-7