Please login to your online account to display your discounted pricing

(S)-N-FMOC-2-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-FMOC-2-Bromophenylalanine
Name Note 95%, (98% e.e.)
CAS 220497-47-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
Formula Weight 466.32
MDL Number MFCD01311743
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361340010

  SDS
thermo fisher scientific
361340010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC361345000

  SDS
thermo fisher scientific
361345000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 466.32
Physical Form Powder
Quantity 1g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Synonym N-(9-Fluorenylmethoxycarbonyl)-2-bromophenyl-L-alanine
MDL Number MFCD01311743
Chemical Name or Material (S)-N-FMOC-2-Bromophenylalanine
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
CAS 220497-47-2
Name Note 95%, (98% e.e.)