Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-N-Diphenylphosphino-N-methyl-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine, (S)-Methyl BoPhoz™

Asymmetric hydrogenation

$71.05 - $71.05

Chemical Identifiers

Chemical Name or Material (S)-N-Diphenylphosphino-N-methyl-[ (R)-2- (diphenylphosphino)ferrocenyl]ethylamine, (S)-Methyl BoPhoz™
CAS 406681-09-2
Molecular Formula C37H35FeNP2
Formula Weight 611.5
MDL Number MFCD05865220
Synonym (S)-Methyl BoPhoz™
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4468577

 
alfa aesar™
4468577
0.1g Each for $71.05
Description & Specifications

Specifications

Formula Weight 611.5
Synonym (S)-Methyl BoPhoz™
Chemical Name or Material (S)-N-Diphenylphosphino-N-methyl-[ (R)-2- (diphenylphosphino)ferrocenyl]ethylamine, (S)-Methyl BoPhoz™
Physical Form Solid
Molecular Formula C37H35FeNP2
MDL Number MFCD05865220
CAS 406681-09-2
Color Yellow to orange
Melting Point 80° to 82°C
Quantity 0.1g
Odor Odorless