Please login to your online account to display your discounted pricing

(S)-(-)-N-BOC-tert-Leucinol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426460050

 View more versions of this product

Catalog No. AC426460050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 217.31
Quantity 5g
Color White
Melting Point 103° to 107°C
CAS Max % 100.0
Assay Percent Range 98%
Specific Rotation -5.5° to -7.9° (20°C, 589nm) (c=1, CHCl3)
Molecular Formula C11H23NO3
MDL Number MFCD01861301
Chemical Name or Material (S)-(-)-N-BOC-tert-Leucinol
Packaging Glass bottle
CAS 153645-26-2
CAS Min % 97.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-(-)-N-BOC-tert-Leucinol
CAS 153645-26-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H23NO3
Formula Weight 217.31
MDL Number MFCD01861301