missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-N-BOC-tert-Leucinol, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 153645-26-2 |
---|---|
Molecular Formula | C11H23NO3 |
Molecular Weight (g/mol) | 217.31 |
MDL Number | MFCD01861301 |
InChI Key | AZHJHZWFJVBGIL-MRVPVSSYSA-N |
Synonym | s---n-boc-tert-leucinol, n-boc-s-+-tert-leucinol, s-tert-butyl 1-hydroxy-3,3-dimethylbutan-2-yl carbamate, tert-butyl n-2s-1-hydroxy-3,3-dimethylbutan-2-yl carbamate, n-boc-l-tert-leucinol, s-n-boc-l-tert-leucinol, s-n-boc-tert-leucinol, s-2-tert-butoxycarbonylamino-3,3-dimethyl-1-butanol |
PubChem CID | 10376058 |
IUPAC Name | tert-butyl N-[(2S)-1-hydroxy-3,3-dimethylbutan-2-yl]carbamate |
SMILES | CC(C)(C)C(CO)NC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC426460010
|
Thermo Scientific Chemicals
426460010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC426460050
|
Thermo Scientific Chemicals
426460050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
153645-26-2 | |
217.31 | |
AZHJHZWFJVBGIL-MRVPVSSYSA-N | |
10376058 | |
CC(C)(C)C(CO)NC(=O)OC(C)(C)C |
C11H23NO3 | |
MFCD01861301 | |
s---n-boc-tert-leucinol, n-boc-s-+-tert-leucinol, s-tert-butyl 1-hydroxy-3,3-dimethylbutan-2-yl carbamate, tert-butyl n-2s-1-hydroxy-3,3-dimethylbutan-2-yl carbamate, n-boc-l-tert-leucinol, s-n-boc-l-tert-leucinol, s-n-boc-tert-leucinol, s-2-tert-butoxycarbonylamino-3,3-dimethyl-1-butanol | |
tert-butyl N-[(2S)-1-hydroxy-3,3-dimethylbutan-2-yl]carbamate |
Specifications
153645-26-2 | |
100.0 | |
97.5% min. (GC) | |
C11H23NO3 | |
1g | |
s---n-boc-tert-leucinol, n-boc-s-+-tert-leucinol, s-tert-butyl 1-hydroxy-3,3-dimethylbutan-2-yl carbamate, tert-butyl n-2s-1-hydroxy-3,3-dimethylbutan-2-yl carbamate, n-boc-l-tert-leucinol, s-n-boc-l-tert-leucinol, s-n-boc-tert-leucinol, s-2-tert-butoxycarbonylamino-3,3-dimethyl-1-butanol | |
CC(C)(C)C(CO)NC(=O)OC(C)(C)C | |
217.31 | |
217.31 | |
98% |
97.5 | |
Authentic | |
Glass bottle | |
MFCD01861301 | |
-5.5° to -7.9° (20°C, 589nm) (c=1, CHCl3) | |
AZHJHZWFJVBGIL-MRVPVSSYSA-N | |
tert-butyl N-[(2S)-1-hydroxy-3,3-dimethylbutan-2-yl]carbamate | |
10376058 | |
Crystalline Powder | |
(S)-(-)-N-BOC-tert-Leucinol |
RUO â Research Use Only