missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-BOC-Propargylglycine, 95%, 98% e.e., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 361805000

 View more versions of this product

Catalog No. AC361805000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(S)-N-BOC-Propargylglycine
63039-48-5
100.0
Authentic
Crystalline Powder
95%
213.23
Off-white to light yellow
500mg
95%, (98% e.e.)
94.0
97.5% min. (GC)
Glass bottle
≥94%
C10H15NO4
MFCD01320855
(S)-N-tert-Butoxycarbonyl-2-amino-4-pentynoic acid

Chemical Identifiers

63039-48-5
100.0
C10H15NO4
MFCD01320855
(S)-N-BOC-Propargylglycine, 95%, (98% e.e.)
94.0
95%
213.23
(S)-N-tert-Butoxycarbonyl-2-amino-4-pentynoic acid
≥94%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA