Please login to your online account to display your discounted pricing

(S)-N-BOC-Propargylglycine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361800010

 View more versions of this product

Catalog No. AC361800010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 213.23
Quantity 1g
Color Off-white to light yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C10H15NO4
CAS 63039-48-5
Enantiomeric Excess 97.5% min. (GC)
Chemical Name or Material (S)-N-BOC-Propargylglycine
Packaging Glass bottle
Name Note 95%, (98% e.e.)
MDL Number MFCD01320855
Synonym (S)-N-tert-Butoxycarbonyl-2-amino-4-pentynoic acid
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-N-BOC-Propargylglycine
Name Note 95%, (98% e.e.)
CAS 63039-48-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H15NO4
Formula Weight 213.23
MDL Number MFCD01320855
Synonym (S)-N-tert-Butoxycarbonyl-2-amino-4-pentynoic acid