missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-BOC-Propargylglycine, 95%, Thermo Scientific™

$488.99 - $488.99

Chemical Identifiers

CAS 63039-48-5
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.23
MDL Number MFCD01320855
InChI Key AMKHAJIFPHJYMH-ZETCQYMHSA-N
Synonym 2s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, 2s-2-tert-butoxycarbonylamino pent-4-ynoic acid, 4-pentynoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, boc-l-propargyl glycine, boc-l-propargylglycine, boc-pra-oh, boc-propargyl-gly-oh, boc-propargylglycine, n-boc-2-propargyl-l-glycine, s-2-tert-butoxycarbonyl amino pent-4-ynoic acid
PubChem CID 2734488
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
SMILES CC(C)(C)OC(=O)NC(CC#C)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC361800010
SDS View Documents Promotion Details
Thermo Fisher Scientific
361800010
1g Glass bottle
Each for $488.99
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

63039-48-5
213.23
AMKHAJIFPHJYMH-ZETCQYMHSA-N
2734488
CC(C)(C)OC(=O)NC(CC#C)C(=O)O
C10H15NO4
MFCD01320855
2s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, 2s-2-tert-butoxycarbonylamino pent-4-ynoic acid, 4-pentynoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, boc-l-propargyl glycine, boc-l-propargylglycine, boc-pra-oh, boc-propargyl-gly-oh, boc-propargylglycine, n-boc-2-propargyl-l-glycine, s-2-tert-butoxycarbonyl amino pent-4-ynoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid

Specifications

63039-48-5
100.0
85.5°C
94% min. (GC)
Authentic
Glass bottle
2s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, 2s-2-tert-butoxycarbonylamino pent-4-ynoic acid, 4-pentynoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, boc-l-propargyl glycine, boc-l-propargylglycine, boc-pra-oh, boc-propargyl-gly-oh, boc-propargylglycine, n-boc-2-propargyl-l-glycine, s-2-tert-butoxycarbonyl amino pent-4-ynoic acid
AMKHAJIFPHJYMH-ZETCQYMHSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
2734488
Crystalline Powder
(S)-N-BOC-Propargylglycine, 95%, (98% e.e.)
94.0
White to Yellow
97.5% min. (GC)
C10H15NO4
MFCD01320855
1g
Solubility in water: soluble.
CC(C)(C)OC(=O)NC(CC#C)C(=O)O
213.23
213.23
95%
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

TSCA : TSCA

SDS
Product Certifications

RUO – Research Use Only