Please login to your online account to display your discounted pricing

(S)-N-BOC-Prolinal, 97%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC368290250

 View more versions of this product

Catalog No. AC368290250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 199.25
Quantity 25g
Specific Rotation Condition −97.5° (20°C c=1,CHCl3)
Density 1.063g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (S)-N-BOC-Prolinal
Assay Percent Range 97%
Recommended Storage Freezer -20°C
Molecular Formula C10H17NO3
MDL Number MFCD00274186
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym N-(tert-Butoxycarbonyl)-L-prolinal
Name Note 97%
CAS 69610-41-9

Specifications

Chemical Name or Material (S)-N-BOC-Prolinal
Name Note 97%
CAS 69610-41-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H17NO3
Formula Weight 199.25
MDL Number MFCD00274186
Synonym N-(tert-Butoxycarbonyl)-L-prolinal
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Freezer -20°C

SDS & Safety Documents