missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-BOC-α-Ethylalanine, 98%, 98% ee, ACROS Organics™

$205.29 - $205.29

Chemical Identifiers

CAS 151171-11-8
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
InChI Key SHZXLTCEPXVCSV-JTQLQIEISA-N
Synonym 2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, boc-l-isovaline, n-1,1-dimethylethyl oxy carbonyl-d-isovaline, s-2-methyl-2-tert-butoxycarbonylamino butyric acid, s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, s-2-tert-butoxycarbonylamino-2-methylbutanoic acid
PubChem CID 14284792
IUPAC Name (2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C)(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441131000
SDS View Documents
Thermo Fisher Scientific
AC441131000
100mg Each for $205.29
 
AC441135000
SDS View Documents
Thermo Fisher Scientific
AC441135000
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

151171-11-8
217.27
2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, boc-l-isovaline, n-1,1-dimethylethyl oxy carbonyl-d-isovaline, s-2-methyl-2-tert-butoxycarbonylamino butyric acid, s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, s-2-tert-butoxycarbonylamino-2-methylbutanoic acid
(2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
C10H19NO4
SHZXLTCEPXVCSV-JTQLQIEISA-N
14284792
CCC(C)(C(=O)O)NC(=O)OC(C)(C)C

Specifications

98.0% min.
Glass Bottle
White to Yellow
100mg
97.5
98%
2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, boc-l-isovaline, n-1,1-dimethylethyl oxy carbonyl-d-isovaline, s-2-methyl-2-tert-butoxycarbonylamino butyric acid, s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, s-2-tert-butoxycarbonylamino-2-methylbutanoic acid
CCC(C)(C(=O)O)NC(=O)OC(C)(C)C
217.27
217.27
98%,98% ee
Authentic
12
104°C to 112°C
151171-11-8
100.0
C10H19NO4
SHZXLTCEPXVCSV-JTQLQIEISA-N
(2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
14284792
Powder
(S)-N-BOC-α-Ethylalanine, 98% ee