Please login to your online account to display your discounted pricing

(S)-N-BOC-Azetidine-2-carboxylic acid, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  361900010

 View more versions of this product

Catalog No. AC361900010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 201.22
Quantity 1g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Synonym (S)-N-tert-Butoxycarbonyl-azetidine-2-carboxylic acid
Enantiomeric Excess 97.5% min. (HPLC)
Chemical Name or Material (S)-N-BOC-Azetidine-2-carboxylic acid
Assay Percent Range 95%
MDL Number MFCD01321011
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Molecular Formula C9H15NO4
CAS 51077-14-6
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-N-BOC-Azetidine-2-carboxylic acid
Name Note 95%, (98% e.e.)
CAS 51077-14-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H15NO4
Formula Weight 201.22
MDL Number MFCD01321011
Synonym (S)-N-tert-Butoxycarbonyl-azetidine-2-carboxylic acid