Please login to your online account to display your discounted pricing

(S)-N-BOC-Azetidine-2-carboxylic acid, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-Azetidine-2-carboxylic acid
Name Note 95%, (98% e.e.)
CAS 51077-14-6
CAS Min % 94.0
CAS Max % 100.0
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C9H15NO4
Formula Weight 201.22g/mol
MDL Number MFCD01321011
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361900010

  SDS
fisher scientific acros
361900010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color White
Chemical Name or Material (S)-N-BOC-Azetidine-2-carboxylic acid
Name Note 95%, (98% e.e.)
CAS 51077-14-6
CAS Min % 94.0
CAS Max % 100.0
Physical Form White powder
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C9H15NO4
Formula Weight 201.22g/mol
MDL Number MFCD01321011
Synonym (S)-N-tert-Butoxycarbonyl-azetidine-2-carboxylic acid
TSCA TSCA