missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-BOC-Allylglycine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC390100050

 View more versions of this product

Catalog No. AC390100050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(S)-N-BOC-Allylglycine
94.0
Viscous Liquid
95%
C10H17NO4
MFCD01320851
+9° to +12° (20°C, 589nm) (C=1, methanol)
Clear pale yellow
90600-20-7
100.0
≥94%
Authentic
215.25
Glass bottle
+10.50° (20°C c=1,methanol)
5g

Chemical Identifiers

90600-20-7
100.0
C10H17NO4
MFCD01320851
≥94%
94.0
95%
215.25
(S)-N-BOC-Allylglycine