Please login to your online account to display your discounted pricing

(S)-N-BOC-Allylglycine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-Allylglycine
CAS 90600-20-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H17NO4
Formula Weight 215.25
MDL Number MFCD01320851
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390100050

  SDS
thermo fisher scientific
AC390100050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390100010

  SDS
fisher scientific acros
AC390100010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390105000

  SDS
thermo fisher scientific
AC390105000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 215.25
Physical Form Viscous Liquid
Quantity 5g
Color Clear pale yellow
Specific Rotation Condition +10.50° (20°C c=1,methanol)
CAS Max % 100.0
CAS Min % 94.0
Specific Rotation +9° to +12° (20°C, 589nm) (C=1, methanol)
Assay Percent Range 95%
Molecular Formula C10H17NO4
Chemical Name or Material (S)-N-BOC-Allylglycine
CAS 90600-20-7
MDL Number MFCD01320851
Packaging Glass bottle
Infrared Spectrum Authentic