Please login to your online account to display your discounted pricing

(S)-N-BOC-Allylglycine, 95%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC390105000

 View more versions of this product

Catalog No. AC390105000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Viscous Liquid
Formula Weight 215.25
Quantity 500mg
Color Clear pale yellow
Specific Rotation Condition +10.50° (20°C c=1,methanol)
CAS Min % 94.0
CAS Max % 100.0
CAS 90600-20-7
Packaging Glass bottle
Assay Percent Range 95%
Chemical Name or Material (S)-N-BOC-Allylglycine
Molecular Formula C10H17NO4
MDL Number MFCD01320851
Specific Rotation +9° to +12° (20°C, 589nm) (C=1, methanol)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-N-BOC-Allylglycine
CAS 90600-20-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H17NO4
Formula Weight 215.25
MDL Number MFCD01320851