Please login to your online account to display your discounted pricing

(S)-N-BOC-4-Thiazoylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361780010

 View more versions of this product

Catalog No. AC361780010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 272.32
Quantity 1g
Color White to pale yellow-beige
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H16N2O4S
Enantiomeric Excess 97.5% min. (HPLC)
Name Note 95%, (98% e.e.)
CAS 119434-75-2
MDL Number MFCD00079682
Chemical Name or Material (S)-N-BOC-4-Thiazoylalanine
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym N-tert-Butoxycarbonyl-4-thiazoyl-L-alanine

Specifications

Chemical Name or Material (S)-N-BOC-4-Thiazoylalanine
Name Note 95%, (98% e.e.)
CAS 119434-75-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H16N2O4S
Formula Weight 272.32
MDL Number MFCD00079682
Synonym N-tert-Butoxycarbonyl-4-thiazoyl-L-alanine