Please login to your online account to display your discounted pricing

(S)-N-BOC-4-Thiazoylalanine, 95%, 98% e.e., ACROS Organics™

$189.75 - $189.75

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-4-Thiazoylalanine
Name Note 95%, (98% e.e.)
CAS 119434-75-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H16N2O4S
Formula Weight 272.32
MDL Number MFCD00079682
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361780010

  SDS
thermo fisher scientific
361780010
Glass bottle 1g Each for $189.75
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 272.32
Quantity 1g
Color White to pale yellow-beige
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H16N2O4S
Enantiomeric Excess 97.5% min. (HPLC)
Name Note 95%, (98% e.e.)
CAS 119434-75-2
MDL Number MFCD00079682
Chemical Name or Material (S)-N-BOC-4-Thiazoylalanine
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym N-tert-Butoxycarbonyl-4-thiazoyl-L-alanine