Please login to your online account to display your discounted pricing

(S)-N-BOC-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 142186-36-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H16FNO4
Formula Weight 269.27
MDL Number MFCD01320862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC361920010

  SDS
thermo fisher scientific
361920010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 269.27
Quantity 1g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C13H16FNO4
MDL Number MFCD01320862
CAS 142186-36-5
Chemical Name or Material (S)-N-BOC-4-Fluorophenylglycine
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Synonym (L)-N-tert-Butoxycarbonyl-α-(4-fluorophenyl)acetic acid