Please login to your online account to display your discounted pricing

(S)-N-BOC-4-Fluorophenylalanine, 95%, 98% e.e., Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  36188-0050

 View more versions of this product

Catalog No. AC361880050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 283.3
Quantity 5g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (S)-N-BOC-4-Fluorophenylalanine
Synonym N-tert-Butoxycarbonyl-4-fluorophenyl-L-alanine
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C14H18FNO4
MDL Number MFCD00079672
CAS 41153-30-4

Specifications

Chemical Name or Material (S)-N-BOC-4-Fluorophenylalanine
Name Note 95%, (98% e.e.)
CAS 41153-30-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H18FNO4
Formula Weight 283.3
MDL Number MFCD00079672
Synonym N-tert-Butoxycarbonyl-4-fluorophenyl-L-alanine