Please login to your online account to display your discounted pricing

(S)-N-BOC-4-Fluorophenylalanine, 95%, 98% e.e., Acros Organics

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-4-Fluorophenylalanine
Name Note 95%, (98% e.e.)
CAS 41153-30-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H18FNO4
Formula Weight 283.3
MDL Number MFCD00079672
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361880050

  SDS
thermo fisher scientific
36188-0050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 283.3
Physical Form Powder
Quantity 5g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Synonym N-tert-Butoxycarbonyl-4-fluorophenyl-L-alanine
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min.
Chemical Name or Material (S)-N-BOC-4-Fluorophenylalanine
Molecular Formula C14H18FNO4
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
MDL Number MFCD00079672
CAS 41153-30-4