Please login to your online account to display your discounted pricing

(S)-N-BOC-4-Chlorophenylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361940050

 View more versions of this product

Catalog No. AC361940050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Granular Powder
Formula Weight 299.75
Quantity 5g
Color Pale yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
MDL Number MFCD00057808
Synonym N-tert-Butoxycarbonyl-4-chlorophenyl-L-alanine
Infrared Spectrum Authentic
CAS 68090-88-0
Chemical Name or Material (S)-N-BOC-4-Chlorophenylalanine
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C14 H18 Cl N O4
Enantiomeric Excess 97.5% min. (HPLC)

Specifications

Chemical Name or Material (S)-N-BOC-4-Chlorophenylalanine
Name Note 95%, (98% e.e.)
CAS 68090-88-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H18 Cl N O4
Formula Weight 299.75
MDL Number MFCD00057808
Synonym N-tert-Butoxycarbonyl-4-chlorophenyl-L-alanine