Please login to your online account to display your discounted pricing

(S)-N-BOC-4-Chlorophenylalanine, 95%, 98% e.e., ACROS Organics™

$203.28 - $203.28

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-4-Chlorophenylalanine
Name Note 95%, (98% e.e.)
CAS 68090-88-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H18 Cl N O4
Formula Weight 299.75
MDL Number MFCD00057808
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361940010

  SDS
thermo fisher scientific
361940010
Glass bottle 1g Each for $203.28

AC361940050

  SDS
thermo fisher scientific
361940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 299.75
Physical Form Granular Powder
Quantity 1g
Color Pale yellow
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Assay Percent Range 95%
CAS 68090-88-0
Name Note 95%, (98% e.e.)
Molecular Formula C14 H18 Cl N O4
MDL Number MFCD00057808
Chemical Name or Material (S)-N-BOC-4-Chlorophenylalanine
Synonym N-tert-Butoxycarbonyl-4-chlorophenyl-L-alanine
Enantiomeric Excess 97.5% min. (HPLC)
Packaging Glass bottle
Infrared Spectrum Authentic