missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-N-BOC-3-pyrrolidinol, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC397580050

 View more versions of this product

Catalog No. AC397580050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-(+)-N-BOC-3-pyrrolidinol
Glass bottle
273.3°C
60.0°C to 64.0°C
101469-92-5
100.0
C9H17NO3
n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-1-boc-3-hydroxypyrrolidine, s-+-n-boc-3-pyrrolidinol, s-1-boc-3-hydroxypyrrolidine, s-1-boc-3-pyrrolidinol, s-1-n-boc-3-hydroxypyrrolidine, s-bochp, s-n-boc-3-pyrrolidinol, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)O
178.24
178.24
98+%
Authentic
+21° to +26° (20°C, 589nm) (C=1, in Methanol)
White to Yellow
5g
98.0
98% min. (HPLC)
MFCD01317839
APCBTRDHCDOPNY-ZETCQYMHSA-N
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
854055
Crystalline Powder

Chemical Identifiers

101469-92-5
178.24
APCBTRDHCDOPNY-ZETCQYMHSA-N
854055
CC(C)(C)OC(=O)N1CCC(C1)O
C9H17NO3
MFCD01317839
n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-1-boc-3-hydroxypyrrolidine, s-+-n-boc-3-pyrrolidinol, s-1-boc-3-hydroxypyrrolidine, s-1-boc-3-pyrrolidinol, s-1-n-boc-3-hydroxypyrrolidine, s-bochp, s-n-boc-3-pyrrolidinol, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents