missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-N-BOC-3-pyrrolidinol, +98%, ACROS Organics™

Manufacturer:  Acros Organics AC397580010

 View more versions of this product

Catalog No. AC397580010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-(+)-N-BOC-3-pyrrolidinol
Glass bottle
273.3°C
60.0°C to 64.0°C
101469-92-5
100.0
C9H17NO3
n-tert-butoxycarbonyl-s-+-3-pyrrolidinol
CC(C)(C)OC(=O)N1CCC(C1)O
178.24
178.24
≥98%
Authentic
+21° to +26° (20°C, 589nm) (C=1, in Methanol)
White to Yellow
1g
98.0
98+%
MFCD01317839
APCBTRDHCDOPNY-ZETCQYMHSA-N
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
854055
Crystalline Powder

Chemical Identifiers

101469-92-5
178.24
APCBTRDHCDOPNY-ZETCQYMHSA-N
854055
CC(C)(C)OC(=O)N1CCC(C1)O
C9H17NO3
MFCD01317839
n-tert-butoxycarbonyl-s-+-3-pyrrolidinol
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents