Please login to your online account to display your discounted pricing

(S)-N-BOC-(3-Pyridyl)alanine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361950010

 View more versions of this product

Catalog No. AC361950010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 266.29
Quantity 1g
Color Off-white to pale yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Molecular Formula C13H18N2O4
Enantiomeric Excess 97.5% min. (HPLC)
Chemical Name or Material (S)-N-BOC-(3-Pyridyl)alanine
Assay Percent Range 95%
CAS 117142-26-4
Name Note 95%, (98% e.e.)
MDL Number MFCD00079681
Synonym N-tert-Butoxycarbonyl-3-pyridyl-L-alanine
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-N-BOC-(3-Pyridyl)alanine
Name Note 95%, (98% e.e.)
CAS 117142-26-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H18N2O4
Formula Weight 266.29
MDL Number MFCD00079681
Synonym N-tert-Butoxycarbonyl-3-pyridyl-L-alanine