Please login to your online account to display your discounted pricing

(S)-N-BOC-(3-Pyridyl)alanine, 95%, 98% e.e., ACROS Organics™

$318.50 - $318.50

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-(3-Pyridyl)alanine
Name Note 95%, (98% e.e.)
CAS 117142-26-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H18N2O4
Formula Weight 266.29
MDL Number MFCD00079681
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361950010

  SDS
thermo fisher scientific
361950010
Glass bottle 1g Each for $318.50

AC361950050

  SDS
thermo fisher scientific
361950050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 266.29
Quantity 1g
Color Off-white to pale yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Molecular Formula C13H18N2O4
Enantiomeric Excess 97.5% min. (HPLC)
Chemical Name or Material (S)-N-BOC-(3-Pyridyl)alanine
Assay Percent Range 95%
CAS 117142-26-4
Name Note 95%, (98% e.e.)
MDL Number MFCD00079681
Synonym N-tert-Butoxycarbonyl-3-pyridyl-L-alanine
Infrared Spectrum Authentic