Please login to your online account to display your discounted pricing

(S)-N-BOC-3-Cyanophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-3-Cyanophenylalanine
Name Note 95%, (98% e.e.)
CAS 131980-30-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15 H18 N2 O4
Formula Weight 290.31
MDL Number MFCD00797560
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361930050

  SDS
thermo fisher scientific
361930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 290.31
Quantity 5g
Color Off-white to light yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%, (98% e.e.)
CAS 131980-30-8
Assay Percent Range 95%
MDL Number MFCD00797560
Chemical Name or Material (S)-N-BOC-3-Cyanophenylalanine
Molecular Formula C15 H18 N2 O4
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
Synonym N-tert-Butoxycarbonyl-3-cyanophenyl-L-alanine
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.