missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-1-BOC-3-aminopyrrolidine, 95%, ACROS Organics™

Chemical Identifiers

CAS 147081-44-5
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.25
InChI Key CMIBWIAICVBURI-ZETCQYMHSA-N
Synonym s---1-boc-3-aminopyrrolidine, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s---n-boc-3-aminopyrrolidine, s-1-boc-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s-3-amino-n-boc-pyrrolidine, s-bocap, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate
PubChem CID 854071
IUPAC Name tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC435450050
SDS View Documents
Thermo Fisher Scientific
AC435450050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

147081-44-5
186.25
s---1-boc-3-aminopyrrolidine, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s---n-boc-3-aminopyrrolidine, s-1-boc-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s-3-amino-n-boc-pyrrolidine, s-bocap, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate
tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
C9H18N2O2
CMIBWIAICVBURI-ZETCQYMHSA-N
854071
CC(C)(C)OC(=O)N1CCC(C1)N

Specifications

91°C
Glass bottle
120.0°C to 125.0°C (5.0 Torr)
5g
94.0
94% min. (GC)
s---1-boc-3-aminopyrrolidine, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s---n-boc-3-aminopyrrolidine, s-1-boc-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s-3-amino-n-boc-pyrrolidine, s-bocap, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)N
186.25
186.25
95%
Authentic
-1.5° to -3.0° (20°C, 589nm) (C=1, chloroform)
Colorless to Yellow
147081-44-5
100.0
C9H18N2O2
CMIBWIAICVBURI-ZETCQYMHSA-N
tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
854071
Liquid
(S)-(-)-1-BOC-3-aminopyrrolidine
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Risk of explosion if heated under confinement.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

SDS