Please login to your online account to display your discounted pricing

(S)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
Name Note 95%, (98% e.e.)
CAS 261165-06-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H18 Br N O4
Formula Weight 344.21
MDL Number MFCD03419292
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361860010

  SDS
thermo fisher scientific
361860010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 344.21
Quantity 1g
Color White to off-white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Enantiomeric Excess 97.5% min.
Chemical Name or Material (S)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
Name Note 95%, (98% e.e.)
Molecular Formula C14 H18 Br N O4
CAS 261165-06-4
MDL Number MFCD03419292
Assay Percent Range 95%
Synonym (S)-N-BOC-β-(4-bromophenyl)-β-alanine
Infrared Spectrum Authentic