Please login to your online account to display your discounted pricing

(S)-N-BOC-2-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361670010

 View more versions of this product

Catalog No. AC361670010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 344.21
Physical Form Crystalline Powder
Quantity 1g
Color Pale yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
CAS 261165-02-0
Synonym N-tert-Butoxycarbonyl-2-bromophenyl-L-alanine
Enantiomeric Excess 97.5% min.
Chemical Name or Material (S)-N-BOC-2-Bromophenylalanine
Assay Percent Range 95%
Molecular Formula C14H18BrNO4
Name Note 95%, (98% e.e.)
MDL Number MFCD01317709
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-N-BOC-2-Bromophenylalanine
Name Note 95%, (98% e.e.)
CAS 261165-02-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H18BrNO4
Formula Weight 344.21
MDL Number MFCD01317709
Synonym N-tert-Butoxycarbonyl-2-bromophenyl-L-alanine