missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-BOC-2-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

CAS 261165-02-0
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C14H18BrNO4
Formula Weight 344.21
MDL Number MFCD01317709
Synonym N-tert-Butoxycarbonyl-2-bromophenyl-L-alanine
Chemical Name or Material (S)-N-BOC-2-Bromophenylalanine, 95%, (98% e.e.)
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC361675000 SDS Thermo Fisher Scientific
361675000
Glass bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC361670010 SDS Thermo Fisher Scientific
361670010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

261165-02-0
100.0
C14H18BrNO4
MFCD01317709
(S)-N-BOC-2-Bromophenylalanine, 95%, (98% e.e.)
94.0
95%
344.21
N-tert-Butoxycarbonyl-2-bromophenyl-L-alanine
≥94%

Specifications

261165-02-0
100.0
Authentic
Crystalline Powder
C14H18BrNO4
MFCD01317709
Pale yellow
(S)-N-BOC-2-Bromophenylalanine, 95%, (98% e.e.)
94.0
97.5% min.
Glass bottle
95%
344.21
500mg
N-tert-Butoxycarbonyl-2-bromophenyl-L-alanine
≥94%

Safety and Handling

TSCA : TSCA