missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-BOC-2-Amino-3-phenylpropyl cyanide, 97+%, ACROS Organics™

Chemical Identifiers

CAS 172695-25-9
Molecular Formula C15H20N2O2
Molecular Weight (g/mol) 260.337
MDL Number MFCD06658950
InChI Key BEJMQXQXIQDKCZ-CYBMUJFWSA-N
Synonym s-tert-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-n-boc-2-amino-3-phenylpropyl cyanide, tert-butyl n-2s-1-cyano-3-phenylpropan-2-yl carbamate, s-t-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-3-tert-butoxycarbonylamino-4-phenylbutanenitrile, tert-butyl s-1-cyano-3-phenylpropan-2-yl carbamate, tert-butyl 2s-1-cyano-3-phenylpropan-2-yl carbamate, 5-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester, s-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester
PubChem CID 2734830
IUPAC Name tert-butyl N-[(2S)-1-cyano-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC#N)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC374520050 SDS View Documents Acros Organics
AC374520050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC374520010 SDS View Documents Acros Organics
AC374520010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

172695-25-9
260.337
BEJMQXQXIQDKCZ-CYBMUJFWSA-N
2734830
CC(C)(C)OC(=O)NC(CC#N)CC1=CC=CC=C1
C15H20N2O2
MFCD06658950
s-tert-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-n-boc-2-amino-3-phenylpropyl cyanide, tert-butyl n-2s-1-cyano-3-phenylpropan-2-yl carbamate, s-t-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-3-tert-butoxycarbonylamino-4-phenylbutanenitrile, tert-butyl s-1-cyano-3-phenylpropan-2-yl carbamate, tert-butyl 2s-1-cyano-3-phenylpropan-2-yl carbamate, 5-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester, s-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester
tert-butyl N-[(2S)-1-cyano-3-phenylpropan-2-yl]carbamate

Specifications

Authentic
White
172695-25-9
100.0
C15H20N2O2
MFCD06658950
BEJMQXQXIQDKCZ-CYBMUJFWSA-N
tert-butyl N-[(2S)-1-cyano-3-phenylpropan-2-yl]carbamate
2734830
Crystalline Powder
(S)-N-BOC-2-Amino-3-phenylpropyl cyanide
Glass bottle
5g
97.0
97+%
C6H5CH2CH(CH2CN)NHCO2C(CH3)2
s-tert-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-n-boc-2-amino-3-phenylpropyl cyanide, tert-butyl n-2s-1-cyano-3-phenylpropan-2-yl carbamate, s-t-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-3-tert-butoxycarbonylamino-4-phenylbutanenitrile, tert-butyl s-1-cyano-3-phenylpropan-2-yl carbamate, tert-butyl 2s-1-cyano-3-phenylpropan-2-yl carbamate, 5-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester, s-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NC(CC#N)CC1=CC=CC=C1
260.337
260.34
≥97.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning