Please login to your online account to display your discounted pricing

(S)-N-BOC-2-Amino-3-phenylpropyl cyanide, 97+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC374520010

 View more versions of this product

Catalog No. AC374520010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Crystalline Powder
Formula Weight 260.34
Quantity 1g
Color White to off-white
CAS Max % 100.0
CAS Min % 97.0
Linear Formula C6H5CH2CH(CH2CN)NHCO2C(CH3)2
Molecular Formula C15H20N2O2
Assay Percent Range 97+%
CAS 172695-25-9
Chemical Name or Material (S)-N-BOC-2-Amino-3-phenylpropyl cyanide
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD06658950
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

Specifications

Chemical Name or Material (S)-N-BOC-2-Amino-3-phenylpropyl cyanide
CAS 172695-25-9
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C15H20N2O2
Linear Formula C6H5CH2CH(CH2CN)NHCO2C(CH3)2
Formula Weight 260.34
MDL Number MFCD06658950
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents