Please login to your online account to display your discounted pricing

(S)-N-BOC-1-Naphthylalanine, 95%, 98% ee, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-BOC-1-Naphthylalanine
Name Note 95%, (98% e.e.)
CAS 55447-00-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C18 H21 N O4
Formula Weight 315.37
MDL Number MFCD00085377
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361980010

  SDS
thermo fisher scientific
361980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 315.37
Quantity 1g
Color Off-white to beige
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Synonym N-tert-Butoxycarbonyl-1-naphthyl-L-alanine
Molecular Formula C18 H21 N O4
Infrared Spectrum Authentic
Chemical Name or Material (S)-N-BOC-1-Naphthylalanine
Assay Percent Range 95%
MDL Number MFCD00085377
Name Note 95%, (98% e.e.)
CAS 55447-00-2
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)