Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%

Chemical Identifiers

Chemical Name or Material (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
CAS 78879-20-6
Formula Weight 277.32
MDL Number MFCD00143845
Beilstein 4297114
Synonym Boc-Tic-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6142303

  SDS
alfa aesar™
H6142303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6142306

  SDS
alfa aesar™
H6142306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6142314

  SDS
alfa aesar™
H6142314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 277.32
Chemical Name or Material (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Physical Form Solid
Assay 98%
MDL Number MFCD00143845
CAS 78879-20-6
Beilstein 4297114
Synonym Boc-Tic-OH
Color White to off-white
Storage Note 1 2° to 8°C
Quantity 1g
Safety and Handling

2° to 8°C