missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ H6142314

 View more versions of this product

Catalog No. AAH6142314



Specifications

Specifications

(S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
25g
C15H19NO4
4297114
HFPVZPNLMJDJFB-LBPRGKRZSA-N
(3S)-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
664088
Solid
White
78879-20-6
MFCD00143845
3s-2-2-methylpropan-2-yl oxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid
CC(C)(C)OC(=O)N1CC2=CC=CC=C2CC1C(=O)O
277.32
277.32
98%

Chemical Identifiers

78879-20-6
277.32
HFPVZPNLMJDJFB-LBPRGKRZSA-N
664088
CC(C)(C)OC(=O)N1CC2=CC=CC=C2CC1C(=O)O
C15H19NO4
MFCD00143845
3s-2-2-methylpropan-2-yl oxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid
(3S)-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents