Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-N-Benzyloxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%

Chemical Identifiers

Chemical Name or Material (S)-N-Benzyloxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
CAS 79261-58-8
Formula Weight 311.34
MDL Number MFCD00672545
Beilstein 7221387
Synonym Z-Tic-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6540806

 
alfa aesar™
H6540806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6540814

 
alfa aesar™
H6540814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 136° to 140°C
Formula Weight 311.34
Chemical Name or Material (S)-N-Benzyloxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Assay 95%
MDL Number MFCD00672545
Physical Form Solid
Beilstein 7221387
Color White to pale yellow
Storage Note 1 2° to 8°C
Synonym Z-Tic-OH
Quantity 5g
CAS 79261-58-8
Safety and Handling

2° to 8°C