Please login to your online account to display your discounted pricing

(S)-(-)-N-Benzyl-α-methylbenzylamine, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  254870100

 View more versions of this product

Catalog No. AC254870100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5620 to 1.5640 (20°C, 589nm)
Specific Rotation -38° to -40° (20°C, 589nm) (neat)
Density 1g/mL
Specific Rotation Condition +39° (20°C neat)
Color Clear colorless to yellow
Quantity 10g
Chemical Name or Material (S)-(-)-N-Benzyl-α-methylbenzylamine
CAS 17480-69-2
CAS Min % 97.5
CAS Max % 100.0
Physical Form Clear colorless to yellow liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C15H17N
Formula Weight 211.31g/mol
MDL Number MFCD00066325
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-(-)-N-Benzyl-α-methylbenzylamine
CAS 17480-69-2
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C15H17N
Formula Weight 211.31g/mol
MDL Number MFCD00066325
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents