Please login to your online account to display your discounted pricing

(S)-(-)-N-Benzyl-α-methylbenzylamine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(-)-N-Benzyl-α-methylbenzylamine
CAS 17480-69-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H17N
Formula Weight 211.31
MDL Number MFCD00066325
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC254870010

  SDS
thermo fisher scientific
254870010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 211.31
Quantity 1g
Color Clear colorless to yellow
CAS Max % 100.0
Assay Percent Range 98%
CAS 17480-69-2
Refractive Index 1.5620 to 1.5640 (20°C, 589nm)
Molecular Formula C15H17N
Chemical Name or Material (S)-(-)-N-Benzyl-α-methylbenzylamine
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00066325
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Packaging Glass bottle
Specific Rotation -38° to -40° (20°C, 589nm) (neat)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.