missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-N-Benzyl-α-methylbenzylamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 254870010

 View more versions of this product

Catalog No. AC254870010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-(-)-N-Benzyl-α-methylbenzylamine
>110°C
Glass bottle
1
129.0°C (3.0 mmHg)
1g
97.5
97.5% min. (GC)
MFCD00066325
ZYZHMSJNPCYUTB-ZDUSSCGKSA-N
(1S)-N-benzyl-1-phenylethanamine
1268085
Liquid
1.0000g/mL
Authentic
1.5620 to 1.5640
− 39.00
Colorless to Yellow
17480-69-2
100.0
C15H17N
benzyl 1s-1-phenylethyl amine, n-benzyl-d-amphetamine, pubchem5973, s---n-benzyl-1-phenylethylamine, s---n-benzyl-alpha-methylbenzylamine, s--n-benzyl-alpha-phenylethylamine, s-n-benzyl-1-phenylethanamine, s-n-benzyl-1-phenylethylamine, s-n-benzyl-alpha-methylbenzylamine
CC(C1=CC=CC=C1)NCC2=CC=CC=C2
211.31
211.31
98%

Chemical Identifiers

17480-69-2
211.31
ZYZHMSJNPCYUTB-ZDUSSCGKSA-N
1268085
CC(C1=CC=CC=C1)NCC2=CC=CC=C2
C15H17N
MFCD00066325
benzyl 1s-1-phenylethyl amine, n-benzyl-d-amphetamine, pubchem5973, s---n-benzyl-1-phenylethylamine, s---n-benzyl-alpha-methylbenzylamine, s--n-benzyl-alpha-phenylethylamine, s-n-benzyl-1-phenylethanamine, s-n-benzyl-1-phenylethylamine, s-n-benzyl-alpha-methylbenzylamine
(1S)-N-benzyl-1-phenylethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents