Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros 99%, ee 99+%

$53.57 - $161.62

Chemical Identifiers

Chemical Name or Material (S)-(-)-N-Benzyl-1-phenylethylamine
CAS 17480-69-2
Molecular Formula C15H17N
Formula Weight 211.31
MDL Number MFCD00066325
Beilstein 3650264
Synonym L-(-)-N-Benzyl-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1929906

  SDS
alfa aesar™
L1929906
5g Each for $53.57

AAL1929914

  SDS
alfa aesar™
L1929914
25g Each for $161.62
Description & Specifications

Specifications

Formula Weight 211.31
Chemical Name or Material (S)-(-)-N-Benzyl-1-phenylethylamine
Molecular Formula C15H17N
CAS 17480-69-2
Beilstein 3650264
Health Hazard 1 Danger
Assay 99%
Assay Percent Notes ee ≥99%
MDL Number MFCD00066325
Synonym L-(-)-N-Benzyl-alpha-methylbenzylamine
Health Hazard 2 GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 171°C (15mmHg)
Quantity 5g
Flash Point >110°C (230°F)
Optical Rotation −39° (Neat)
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.