missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-N-(5-Nitro-2-pyridyl)prolinol, TCI America™

$308.63 - $1,032.80

Chemical Identifiers

CAS 88374-37-2
Molecular Formula C10H13N3O3
Molecular Weight (g/mol) 223.232
MDL Number MFCD00145250
InChI Key YBARIBJRNJYVTK-VIFPVBQESA-N
Synonym (S)-(-)-1-(5-Nitro-2-pyridyl)-2-pyrrolidinemethanol, 2-(N-Prolinol)-5-nitropyridine
PubChem CID 44630404
IUPAC Name [(2S)-1-(5-nitropyridin-2-yl)pyrrolidin-2-yl]methanol
SMILES C1CC(N(C1)C2=NC=C(C=C2)[N+](=O)[O-])CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N05405G View Documents TCI America
N05405G
5g Each for $1,032.80
 
N05401G View Documents TCI America
N05401G
1g Each for $308.63
 
Specifications

Chemical Identifiers

88374-37-2
223.232
YBARIBJRNJYVTK-VIFPVBQESA-N
44630404
C1CC(N(C1)C2=NC=C(C=C2)[N+](=O)[O-])CO
C10H13N3O3
MFCD00145250
(S)-(-)-1-(5-Nitro-2-pyridyl)-2-pyrrolidinemethanol, 2-(N-Prolinol)-5-nitropyridine
[(2S)-1-(5-nitropyridin-2-yl)pyrrolidin-2-yl]methanol

Specifications

Yellow
5g
C10H13N3O3
(S)-(-)-1-(5-Nitro-2-pyridyl)-2-pyrrolidinemethanol, 2-(N-Prolinol)-5-nitropyridine
C1CC(N(C1)C2=NC=C(C=C2)[N+](=O)[O-])CO
223.232
223.23
≥99.0% (T)
83°C
88374-37-2
MFCD00145250
YBARIBJRNJYVTK-VIFPVBQESA-N
[(2S)-1-(5-nitropyridin-2-yl)pyrrolidin-2-yl]methanol
44630404
Crystal-Powder at 20°C
(S)-(-)-N-(5-Nitro-2-pyridyl)prolinol
Safety and Handling

Safety and Handling

TSCA : No