missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-N-(3-Pentyl)-1-phenylethylamine hydrochloride, 99%

$89.45 - $325.39

Chemical Identifiers

CAS 374790-92-8
Molecular Formula C13H22ClN
Molecular Weight (g/mol) 227.776
MDL Number MFCD06199157
InChI Key RKPRPVRHVBFULB-MERQFXBCSA-N
Synonym s---n-3-pentyl-1-phenylethylamine hydrochloride, s-n-1-phenylethyl pentan-3-amine hydrochloride, pentan-3-yl 1s-1-phenylethyl amine hydrochloride
PubChem CID 22831494
IUPAC Name N-[(1S)-1-phenylethyl]pentan-3-amine;hydrochloride
SMILES CCC(CC)NC(C)C1=CC=CC=C1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1835706 View Documents Alfa Aesar™
L1835706
5g Each for $325.39
 
AAL1835703 View Documents Alfa Aesar™
L1835703
1g Each for $89.45
 
Specifications

Chemical Identifiers

374790-92-8
227.776
RKPRPVRHVBFULB-MERQFXBCSA-N
22831494
CCC(CC)NC(C)C1=CC=CC=C1.Cl
C13H22ClN
MFCD06199157
s---n-3-pentyl-1-phenylethylamine hydrochloride, s-n-1-phenylethyl pentan-3-amine hydrochloride, pentan-3-yl 1s-1-phenylethyl amine hydrochloride
N-[(1S)-1-phenylethyl]pentan-3-amine;hydrochloride

Specifications

232°C to 234°C
374790-92-8
MFCD06199157
RKPRPVRHVBFULB-MERQFXBCSA-N
N-[(1S)-1-phenylethyl]pentan-3-amine;hydrochloride
22831494
99%
5g
C13H22ClN
s---n-3-pentyl-1-phenylethylamine hydrochloride, s-n-1-phenylethyl pentan-3-amine hydrochloride, pentan-3-yl 1s-1-phenylethyl amine hydrochloride
CCC(CC)NC(C)C1=CC=CC=C1.Cl
227.776
227.78
(S)-(-)-N-(3-Pentyl)-1-phenylethylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning