missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-3-Cyanophenylalanine, 95%, 98% e.e., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 362060010

 View more versions of this product

Catalog No. AC362060010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
190.2
White to slightly yellow
100.0
Glass bottle
MFCD01860885
57213-48-6
C10H10N2O2
Powder
1g
94.0
95%
97.5% min. (HPLC)
95%, (98% e.e.)
(S)-N-3-Cyanophenylalanine
Authentic

Chemical Identifiers

57213-48-6
100.0
C10H10N2O2
MFCD01860885
≥94%
94.0
95%
190.2
(S)-N-3-Cyanophenylalanine, 95%, (98% e.e.)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA