Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine, 98%

$56.05 - $700.65

Chemical Identifiers

Chemical Name or Material (S)-(+)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
CAS 69632-31-1
Molecular Formula C15H13N3O5
Formula Weight 315.28
MDL Number MFCD00064498
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1665503

 
alfa aesar™
A1665503
1g Each for $56.05

AAA1665506

 
alfa aesar™
A1665506
5g Each for $162.73

AAA1665514

 
alfa aesar™
A1665514
25g Each for $700.65
Description & Specifications

Specifications

Formula Weight 315.28
CAS 69632-31-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (S)-(+)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
Molecular Formula C15H13N3O5
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00064498
Health Hazard 1 Warning
Melting Point 158° to 161°C
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.