Please login to your online account to display your discounted pricing

(S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338830100

 View more versions of this product

Catalog No. AC338830100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 165.23
Quantity 10g
Color Clear colorless
TSCA TSCA
Specific Rotation Condition − 44.00 (22.00°C c=1,C2H5OH)
CAS Max % 100.0
Specific Rotation -42.5° to -45.5° (22°C, 589nm) (c=1, C2H5OH)
Synonym (S)-(-)-2-[(1-Phenylethyl)-amino]-ethanol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5320 to 1.5350 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Linear Formula C6H5CH(CH3)NH(CH2)2OH
CAS 66849-29-4
Molecular Formula C10H15NO
Chemical Name or Material (S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine
MDL Number MFCD01862172
Assay Percent Range 99%
Name Note 99%

Specifications

Chemical Name or Material (S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine
Name Note 99%
CAS 66849-29-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H15NO
Linear Formula C6H5CH(CH3)NH(CH2)2OH
Formula Weight 165.23
MDL Number MFCD01862172
Synonym (S)-(-)-2-[(1-Phenylethyl)-amino]-ethanol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents