missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide, TCI America™

$175.70 - $640.16

Chemical Identifiers

CAS 96293-17-3
Molecular Formula C25H24N2O2
Molecular Weight (g/mol) 384.479
MDL Number MFCD00145260
InChI Key IPSABLMEYFYEHS-QHCPKHFHSA-N
PubChem CID 980835
IUPAC Name (2S)-N-(2-benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
SMILES C1CC(N(C1)CC2=CC=CC=C2)C(=O)NC3=CC=CC=C3C(=O)C4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B55195G View Documents TCI America
B55195G
5g Each for $640.16
 
B55191G View Documents TCI America
B55191G
1g Each for $175.70
 
Specifications

Chemical Identifiers

96293-17-3
384.479
IPSABLMEYFYEHS-QHCPKHFHSA-N
(2S)-N-(2-benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
C25H24N2O2
MFCD00145260
980835
C1CC(N(C1)CC2=CC=CC=C2)C(=O)NC3=CC=CC=C3C(=O)C4=CC=CC=C4

Specifications

White-Yellow
5g
C25H24N2O2
IPSABLMEYFYEHS-QHCPKHFHSA-N
(2S)-N-(2-benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
980835
Crystal-Powder at 20°C
(S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
100°C
96293-17-3
MFCD00145260
C1CC(N(C1)CC2=CC=CC=C2)C(=O)NC3=CC=CC=C3C(=O)C4=CC=CC=C4
384.479
384.48
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No