missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-alpha-Methylphenylalanine hydrate, 98%, 98% ee, ACROS Organics™

Chemical Identifiers

CAS 23239-35-2
Molecular Formula C10H13NO2·xH2O
Molecular Weight (g/mol) 179.22
InChI Key HYOWVAAEQCNGLE-JTQLQIEISA-N
Synonym 2s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-benzyl-l-ala, alpha-methyl-l-phe, alpha-methyl-l-phenylalanine, alpha-methylphenylalanine, l-alpha-methylphenylalanine, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropionic acid, s-alpha-methylphenylalanine
PubChem CID 2724754
IUPAC Name (2S)-2-amino-2-methyl-3-phenylpropanoic acid
SMILES CC(CC1=CC=CC=C1)(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC275431000
SDS View Documents
Thermo Fisher Scientific
AC275431000
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC275435000
SDS View Documents
Thermo Fisher Scientific
AC275435000
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

23239-35-2
179.22
2s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-benzyl-l-ala, alpha-methyl-l-phe, alpha-methyl-l-phenylalanine, alpha-methylphenylalanine, l-alpha-methylphenylalanine, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropionic acid, s-alpha-methylphenylalanine
(2S)-2-amino-2-methyl-3-phenylpropanoic acid
C10H13NO2·xH2O
HYOWVAAEQCNGLE-JTQLQIEISA-N
2724754
CC(CC1=CC=CC=C1)(C(=O)O)N

Specifications

(HPLC)> 99.0%
− 18.00
100mg
97.5
98.5% min. (HPLC)
2s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-benzyl-l-ala, alpha-methyl-l-phe, alpha-methyl-l-phenylalanine, alpha-methylphenylalanine, l-alpha-methylphenylalanine, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropionic acid, s-alpha-methylphenylalanine
CC(CC1=CC=CC=C1)(C(=O)O)N
179.22
179.22
98%,98% ee
Authentic
White
23239-35-2
100.0
C10H13NO2·xH2O
HYOWVAAEQCNGLE-JTQLQIEISA-N
(2S)-2-amino-2-methyl-3-phenylpropanoic acid
2724754
Fine Crystalline Powder
(S)-α-Methylphenylalanine hydrate, 99%, (99+% e.e.)
Safety and Handling

Safety and Handling

TSCA : TSCA